Assay Details
| Gene Symbol: Skp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106416.2 | NP_001099886.1 | 3 - 4 | 481 | 60 | |
| GenBank mRNA | BC166459.1 | - | 3 - 4 | 481 | 9027296 | 60 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
G1/S transition of mitotic cell cycle
G2/M transition of mitotic cell cycle
protein polyubiquitination
ubiquitin-dependent protein catabolic process
phosphorylation
positive regulation of intracellular estrogen receptor signaling pathway
regulation of apoptotic process
positive regulation of smooth muscle cell proliferation
regulation of cell cycle
cellular response to cell-matrix adhesion
positive regulation of protein polyubiquitination
ubiquitin-protein transferase activity
protein binding
kinase activity
identical protein binding
ubiquitin protein ligase activity
G1/S transition of mitotic cell cycle
G2/M transition of mitotic cell cycle
protein polyubiquitination
ubiquitin-dependent protein catabolic process
phosphorylation
positive regulation of intracellular estrogen receptor signaling pathway
regulation of apoptotic process
positive regulation of smooth muscle cell proliferation
regulation of cell cycle
cellular response to cell-matrix adhesion
positive regulation of protein polyubiquitination
ubiquitin-protein transferase activity
protein binding
kinase activity
identical protein binding
ubiquitin protein ligase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P20266 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P586883 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 94323 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 817715 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 63914 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 861287 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 707180 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 696737 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 298693 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 763150 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 39159 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 954431 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 105354 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 802133 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 42713 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 52886 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 352487 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 565197 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 83461 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 636764 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 465347 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 277141 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 167686 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 55162 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 153466 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 106983 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P586883 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P20266 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P586883 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P20266 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P20266 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P586883 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P20266 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 49846 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 10471 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10821841 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10821847 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10821846 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10821849 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10821850 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10821845 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10821848 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10821844 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10821842 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10821840 | - | - |
| Rat Genome 230 2.0 Array | 1390823_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1390823_at | - | - |
| Affymetrix HT Rat Focus Array | 1390823_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1390823_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0200021393.rn.1 | TC0200003055.rn.1 | RTA TC |
| Rat Expression Set 230 | 1390823_at | - | - |
| Rat Expression Set 230 | 1390823_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10821840 | - | - |
| Rat Genome U34 Set | rc_AA924839_at | - | - |
| Rat Genome U34 Set | rc_AI172562_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22176264 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1371149 | - | - |

Skp2
Genomic Map