Assay Details
| Gene Symbol: Capsl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106417.1 | NP_001099887.1 | 1 - 2 | 136 | 93 | |
| XM_008760763.1 | XP_008758985.1 | 1 - 2 | 112 | 93 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001106417.1, XM_008760763.1 transcripts.
Set Membership: |
|

Capsl
Genomic Map