Assay Details
| Gene Symbol: Nat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037315.1 | NP_001032392.1 | 2 - 3 | 319 | 130 | |
| GenBank mRNA | U01343.1 | - | 2 - 3 | 319 | 130 | |
| U01345.1 | - | 2 - 3 | 374 | 130 | ||
| U17260.1 | - | 1 - 2 | 139 | 130 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001037315.1, U01343.1, U01345.1, U17260.1 transcripts.
Set Membership: |
|

Nat1
Genomic Map