Assay Details
Gene Symbol: Glb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108192.1 | NP_001101662.1 | 4 - 5 | 539 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P347368 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P665941 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 98022 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 67889 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 965493 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 210267 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 957544 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 705039 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 458782 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 57856 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 143221 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 723716 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 458413 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 387292 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 706603 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 387833 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 181800 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 234632 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 201019 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 843954 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 941152 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 324003 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 524238 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 642669 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 576640 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 577548 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 90032 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 83380 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 427168 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 424079 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 707223 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 544115 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 97578 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504655 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 370421 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 439075 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 851003 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 817692 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 623555 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 122590 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 290537 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 923032 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 550991 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 928602 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 793033 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 266063 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 221052 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 493146 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 675474 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 270961 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 217595 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 423799 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 959327 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 649861 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 829505 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 152244 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 140084 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 142583 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 15803 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 57756 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 11177 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 132166 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 74636 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P665941 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P347368 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P347368 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P665941 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P347368 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P665941 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P347368 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 85717 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 34368 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10913979 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10913981 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10913982 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10913988 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10913976 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10913983 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10913990 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10913984 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10913989 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10913986 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10913978 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10913985 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10913980 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10913977 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10913975 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10913987 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10913974 | - | - |
Rat Genome 230 2.0 Array | 1371727_PM_at | - | - |
Rat Genome 230 2.0 Array | 1371727_at | - | - |
Affymetrix HT Rat Focus Array | 1371727_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1371727_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0800013135.rn.1 | TC0800001652.rn.1 | RTA TC |
Rat Expression Set 230 | 1371727_at | - | - |
Rat Expression Set 230 | 1371727_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10913974 | - | - |
Rat Genome U34 Set | rc_AI045074_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22257561 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1371130 | - | - |
