Assay Details
| Gene Symbol: Eif1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106867.1 | NP_001100337.1 | 2 - 3 | 374 | 85 | |
| GenBank mRNA | BC169125.1 | - | 2 - 3 | 294 | 6919533 | 85 |
| FQ211778.1 | - | 2 - 3 | 377 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Eif1b
Genomic Map