Assay Details
Gene Symbol: Sh3bgrl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001137647.1 | NP_001131119.1 | 1 - 2 | 189 | 58 |
Gene Symbol: LOC102554622 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006243468.2 | XP_006243530.1 | 1 - 2 | 184 | 58 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001137647.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
