Assay Details
Gene Symbol: Sh3bgrl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001137647.1 | NP_001131119.1 | 2 - 3 | 383 | 129 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001137647.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P165986 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 919403 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 258861 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 334235 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 152997 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 314216 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 956745 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 42338 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 654938 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 162046 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 171013 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 39270 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 69477 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 899848 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 574719 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 824123 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 699397 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 586812 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 256879 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 425754 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 538383 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 355263 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 192313 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 892522 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 404994 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 580376 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 562553 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 624286 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 854814 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 530477 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 24539 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 437311 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 476215 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 727816 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 198545 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 45031 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 264847 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 782612 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 52217 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 66276 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 545716 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 806048 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 543720 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 715364 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 881269 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 959841 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 835317 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 375977 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 292625 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 944820 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 691603 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 382410 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 877496 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 331724 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 283824 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 284242 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 627945 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 97964 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 460404 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 677586 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 6081 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 1848 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 330852 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 512441 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 10580 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 60632 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 114002 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 151173 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 164753 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 14192 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 105613 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 17140 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 99738 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 143058 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 176843 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 63262 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 158139 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P165986 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P165986 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P165986 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P119575 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10911937 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10911939 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10911936 | - | - |
Rat Genome 230 2.0 Array | 1389640_PM_at | - | - |
Rat Genome 230 2.0 Array | 1389640_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0800008881.rn.1 | TC0800001215.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0800008883.rn.1 | TC0800001215.rn.1 | RTA TC |
Rat Expression Set 230 | 1389640_PM_at | - | - |
Rat Expression Set 230 | 1389640_at | - | - |
Rat Genome U34 Set | rc_AI030271_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1367663 | - | - |
