Assay Details
| Gene Symbol: Igll1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190341.1 | NP_001177270.1 | 1 - 2 | 191 | 66 | |
| XM_006248656.2 | XP_006248718.1 | 1 - 2 | 201 | 66 | ||
| GenBank mRNA | Z68145.1 | - | 1 - 2 | 85 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Igll1
Genomic Map