Assay Details
| Gene Symbol: Igll1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190341.1 | NP_001177270.1 | 2 - 3 | 308 | 87 | |
| GenBank mRNA | Z68145.1 | - | 2 - 3 | 202 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Igll1
Genomic Map