Assay Details
| Gene Symbol: Prodh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135778.1 | NP_001129250.1 | 1 - 2 | 293 | 64 | |
| XM_006248642.2 | XP_006248704.2 | 1 - 2 | 308 | 64 | ||
| XM_006248644.2 | XP_006248706.2 | 1 - 2 | 308 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prodh
Genomic Map