Assay Details
Gene Symbol: Tmem207 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002727912.4 | XP_002727958.1 | 3 - 4 | 346 | 111 | |
XM_006248530.2 | XP_006248592.1 | 4 - 5 | 406 | 171 |

More Information
Set Membership: |
![]() ![]() ![]() |
