Assay Details
| Gene Symbol: Tmem207 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002727912.4 | XP_002727958.1 | 4 - 5 | 499 | 90 | |
| XM_006248530.2 | XP_006248592.1 | 5 - 6 | 559 | 90 | ||
More Information
Set Membership: |
|

Tmem207
Genomic Map