Assay Details
Gene Symbol: Eif2b5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138866.2 | NP_620221.2 | 2 - 3 | 340 | 61 | |
GenBank mRNA | BC085698.1 | - | 2 - 3 | 340 | 7193458 | 61 |
U19516.1 | - | 2 - 3 | 338 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
