Assay Details
| Gene Symbol: Mrpl40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024865.1 | NP_001020036.1 | 1 - 2 | 133 | 96 | |
| GenBank mRNA | BC097472.1 | - | 1 - 2 | 133 | 7457772 | 96 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mrpl40
Genomic Map