Assay Details
| Gene Symbol: Paip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008772044.1 | XP_008770266.1 | 1 - 2 | 149 | 73 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008772044.1 transcripts.
Set Membership: |
|

Paip2
Genomic Map