Assay Details
| Gene Symbol: Nr3c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012576.2 | NP_036708.2 | 5 - 6 | 1954 | 68 | |
| XM_008772065.1 | XP_008770287.1 | 5 - 6 | 1955 | 68 | ||
| XM_008772066.1 | XP_008770288.1 | 6 - 7 | 2005 | 68 | ||
| GenBank mRNA | AB115420.1 | - | 5 - 6 | 1941 | 68 | |
| AF455050.1 | - | 5 - 6 | 1945 | 68 | ||
| AY066016.2 | - | 5 - 6 | 1949 | 68 | ||
| CB699574.1 | - | 1 - 2 | 156 | 68 | ||
| M14053.1 | - | 5 - 6 | 2012 | 68 | ||
| Y12264.1 | - | 5 - 6 | 1966 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nr3c1
Genomic Map