Assay Details
| Gene Symbol: Epb41l4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107397.2 | NP_001100867.1 | 7 - 8 | 653 | 67 | |
| XM_006254582.2 | XP_006254644.1 | 12 - 13 | 3925 | 67 | ||
| XM_006254583.2 | XP_006254645.1 | 12 - 13 | 3031 | 67 | ||
| XM_006254584.2 | XP_006254646.1 | 13 - 14 | 2677 | 67 | ||
| XM_006254588.2 | XP_006254650.1 | 12 - 13 | 1231 | 67 | ||
| XM_008772040.1 | XP_008770262.1 | 12 - 13 | 1008 | 67 | ||
| GenBank mRNA | BC168858.1 | - | 7 - 8 | 653 | 7097125 | 67 |
More Information
Updated Mapping Information
Set Membership: |
|

Epb41l4a
Genomic Map