Assay Details
| Gene Symbol: Ino80c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017446.1 | NP_001017446.1 | 3 - 4 | 470 | 85 | |
| GenBank mRNA | BC091281.1 | - | 3 - 4 | 470 | 7192011 | 85 |
More Information
Updated Mapping Information
Set Membership: |
|

Ino80c
Genomic Map