Assay Details
| Gene Symbol: Stard4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106159.2 | NP_001099629.1 | 1 - 2 | 164 | 62 | |
| GenBank mRNA | CB547156.1 | - | 1 - 2 | 111 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Stard4
Genomic Map