Assay Details
Gene Symbol: Rbm27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006254677.2 | XP_006254739.1 | 12 - 13 | 2096 | 70 | |
XM_006254678.2 | XP_006254740.1 | 11 - 12 | 1931 | 70 | ||
XM_006254679.2 | XP_006254741.1 | 11 - 12 | 1931 | 70 | ||
XM_006254680.2 | XP_006254742.1 | 11 - 12 | 1802 | 70 | ||
XM_008772179.1 | XP_008770401.1 | 12 - 13 | 2096 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
