Assay Details
| Gene Symbol: Pcdhac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199509.1 | NP_955803.1 | 1 | 331 | 98 | |
| GenBank mRNA | AB113397.1 | - | 1 | 331 | 98 | |
| AY573984.1 | - | 1 | 331 | 98 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Pcdhac1
Genomic Map