Assay Details
| Gene Symbol: Sh3bgr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127682.1 | NP_001121154.1 | 2 - 3 | 373 | 106 | |
| XM_006248184.2 | XP_006248246.1 | 2 - 3 | 435 | 106 | ||
| GenBank mRNA | BC159440.1 | - | 2 - 3 | 337 | 106 | |
| FQ214709.1 | - | 2 - 3 | 231 | 106 | ||
| FQ215323.1 | - | 2 - 3 | 341 | 106 | ||
| FQ216161.1 | - | 2 - 3 | 390 | 106 | ||
| FQ216183.1 | - | 2 - 3 | 240 | 106 | ||
| FQ216585.1 | - | 2 - 3 | 238 | 106 | ||
| FQ216591.1 | - | 2 - 3 | 238 | 106 | ||
| FQ216690.1 | - | 2 - 3 | 290 | 106 | ||
| Gene Symbol: LOC102553613 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006248190.2 | XP_006248252.1 | 2 - 3 | 419 | 106 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Sh3bgr
Genomic Map