Assay Details
| Gene Symbol: Sh3bgr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127682.1 | NP_001121154.1 | 3 - 4 | 459 | 126 | |
| XM_006248184.2 | XP_006248246.1 | 3 - 4 | 521 | 126 | ||
| GenBank mRNA | BC159440.1 | - | 3 - 4 | 423 | 126 | |
| FQ214709.1 | - | 3 - 4 | 317 | 126 | ||
| FQ215323.1 | - | 3 - 4 | 427 | 126 | ||
| FQ216161.1 | - | 3 - 4 | 476 | 126 | ||
| FQ216585.1 | - | 3 - 4 | 324 | 126 | ||
| FQ216591.1 | - | 3 - 4 | 324 | 126 | ||
| FQ216690.1 | - | 3 - 4 | 376 | 126 | ||
| FQ223676.1 | - | 3 - 4 | 377 | 126 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sh3bgr
Genomic Map