Assay Details
| Gene Symbol: Wrb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199373.1 | NP_955405.1 | 2 - 3 | 336 | 91 | |
| GenBank mRNA | BC062049.1 | - | 2 - 3 | 336 | 5598163 | 91 |
More Information
Updated Mapping Information
Set Membership: |
|

Wrb
Genomic Map