Assay Details
| Gene Symbol: LOC100911295 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003751029.3 | XP_003751077.1 | 2 - 3 | 196 | 116 | |
| Gene Symbol: Hmgn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013184.1 | NP_001013202.1 | 2 - 3 | 125 | 116 | |
| GenBank mRNA | BC086423.1 | - | 2 - 3 | 125 | 7302011 | 116 |
| FQ222121.1 | - | 2 - 3 | 164 | 116 | ||
| FQ230156.1 | - | 2 - 3 | 128 | 116 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100911295
Genomic Map