Assay Details
| Gene Symbol: Ttc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108315.1 | NP_001101785.1 | 34 - 35 | 4649 | 67 | |
| XM_008768582.1 | XP_008766804.1 | 34 - 35 | 4644 | 67 | ||
| GenBank mRNA | FQ233738.1 | - | 2 - 3 | 823 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ttc3
Genomic Map