Assay Details
Gene Symbol: Odf3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006242237.1 | XP_006242299.1 | 1 - 2 | 221 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
