Assay Details
| Gene Symbol: Chkb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017177.1 | NP_058873.1 | 6 - 7 | 1016 | 67 | |
| XM_006242181.2 | XP_006242243.2 | 6 - 7 | 1682 | 67 | ||
| XM_008765677.1 | XP_008763899.1 | 6 - 7 | 1679 | 67 | ||
| GenBank mRNA | AB006607.1 | - | 6 - 7 | 1016 | 67 | |
| BC060515.1 | - | 6 - 7 | 992 | 5598184 | 67 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Chkb
Genomic Map