Assay Details
| Gene Symbol: Tymp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012122.1 | NP_001012122.1 | 6 - 7 | 805 | 53 | |
| XM_006242209.2 | XP_006242271.1 | 6 - 7 | 1072 | 53 | ||
| XM_008765730.1 | XP_008763952.1 | 6 - 7 | 1078 | 53 | ||
| XM_008765731.1 | XP_008763953.1 | 6 - 7 | 1078 | 53 | ||
| XM_008765732.1 | XP_008763954.1 | 6 - 7 | 1072 | 53 | ||
| XM_008765733.1 | XP_008763955.1 | 6 - 7 | 1030 | 53 | ||
| GenBank mRNA | BC089830.1 | - | 6 - 7 | 805 | 7323197 | 53 |
More Information
Updated Mapping Information
Set Membership: |
|

Tymp
Genomic Map