Assay Details
Gene Symbol: Tymp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012122.1 | NP_001012122.1 | 7 - 8 | 973 | 55 | |
XM_008765731.1 | XP_008763953.1 | 7 - 8 | 1246 | 55 | ||
XM_008765732.1 | XP_008763954.1 | 7 - 8 | 1240 | 55 | ||
GenBank mRNA | BC089830.1 | - | 7 - 8 | 973 | 7323197 | 55 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
