Assay Details
Gene Symbol: Tymp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012122.1 | NP_001012122.1 | 9 - 10 | 1346 | 124 | |
XM_006242209.2 | XP_006242271.1 | 9 - 10 | 1642 | 124 | ||
XM_008765730.1 | XP_008763952.1 | 9 - 10 | 1648 | 124 | ||
XM_008765731.1 | XP_008763953.1 | 9 - 10 | 1645 | 124 | ||
XM_008765732.1 | XP_008763954.1 | 9 - 10 | 1639 | 124 | ||
XM_008765733.1 | XP_008763955.1 | 9 - 10 | 1600 | 124 | ||
GenBank mRNA | BC089830.1 | - | 9 - 10 | 1346 | 7323197 | 124 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
