Assay Details
| Gene Symbol: Dennd6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108749.1 | NP_001102219.1 | 3 - 4 | 337 | 64 | |
| XM_006242222.2 | XP_006242284.1 | 3 - 4 | 1918 | 64 | ||
| XM_006242223.2 | XP_006242285.1 | 3 - 4 | 1918 | 64 | ||
| XM_006242224.2 | XP_006242286.1 | 104 | 64 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006242224.2 transcripts.
Set Membership: |
|

Dennd6b
Genomic Map