Assay Details
Gene Symbol: Trabd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106788.1 | NP_001100258.1 | 5 - 6 | 527 | 64 | |
XM_006242182.2 | XP_006242244.1 | 5 - 6 | 1089 | 64 | ||
XM_006242183.2 | XP_006242245.1 | 5 - 6 | 942 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
