Assay Details
| Gene Symbol: Amd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031011.3 | NP_112273.3 | 3 - 4 | 642 | 154 | |
| GenBank mRNA | AY724509.1 | - | 3 - 4 | 325 | 154 | |
| BC061532.1 | - | 3 - 4 | 645 | 6921099 | 154 | |
| FM031711.1 | - | 3 - 4 | 289 | 154 | ||
| FQ232080.1 | - | 3 - 4 | 462 | 154 | ||
| FQ234346.1 | - | 3 - 4 | 465 | 154 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Amd1
Genomic Map