Assay Details
| Gene Symbol: Amd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031011.3 | NP_112273.3 | 5 - 6 | 798 | 95 | |
| GenBank mRNA | AY724509.1 | - | 5 - 6 | 481 | 95 | |
| BC061532.1 | - | 5 - 6 | 801 | 6921099 | 95 | |
| FM031711.1 | - | 5 - 6 | 445 | 95 | ||
| FQ232080.1 | - | 5 - 6 | 618 | 95 | ||
| FQ234346.1 | - | 5 - 6 | 621 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Amd1
Genomic Map