Assay Details
| Gene Symbol: Idi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053539.2 | NP_445991.2 | 3 - 4 | 641 | 120 | |
| GenBank mRNA | AF003835.1 | - | 3 - 4 | 622 | 120 | |
| BC089786.1 | - | 3 - 4 | 641 | 7322086 | 120 | |
| FQ219635.1 | - | 3 - 4 | 290 | 120 | ||
| FQ220448.1 | - | 3 - 4 | 288 | 120 | ||
| FQ226929.1 | - | 3 - 4 | 294 | 120 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Idi1
Genomic Map