Assay Details
Gene Symbol: Plk5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170557.1 | NP_001164028.1 | 10 - 11 | 1224 | 73 | |
XM_006240921.2 | XP_006240983.1 | 10 - 11 | 1178 | 73 | ||
XM_006240922.2 | XP_006240984.1 | 10 - 11 | 1178 | 73 | ||
XM_006240923.2 | XP_006240985.1 | 9 - 10 | 1061 | 73 | ||
XM_006240925.2 | XP_006240987.1 | 10 - 11 | 1178 | 73 | ||
XM_006240926.2 | XP_006240988.1 | 10 - 11 | 1178 | 73 | ||
XM_008765098.1 | XP_008763320.1 | 10 - 11 | 1178 | 73 | ||
XM_008765099.1 | XP_008763321.1 | 11 - 12 | 1150 | 73 | ||
XM_008765100.1 | XP_008763322.1 | 10 - 11 | 1308 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
