Assay Details
Gene Symbol: Cactin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081447.1 | NP_001074916.1 | 1 - 2 | 224 | 88 | |
XM_008765126.1 | XP_008763348.1 | 1 - 2 | 715 | 88 | ||
GenBank mRNA | AY934505.1 | - | 1 - 2 | 224 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
