Assay Details
| Gene Symbol: Prss57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003956.1 | NP_001003956.1 | 2 - 3 | 236 | 109 | |
| XM_006240979.2 | XP_006241041.1 | 2 - 3 | 379 | 109 | ||
| XM_008765175.1 | XP_008763397.1 | 2 - 3 | 379 | 109 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prss57
Genomic Map