Assay Details
| Gene Symbol: Cyp4f17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006241085.2 | XP_006241147.1 | 1154 | 145 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006241085.2 transcripts.
Set Membership: |
|

Cyp4f17
Genomic Map