Assay Details
| Gene Symbol: Cyp4f17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191986.1 | NP_001178915.1 | 397 | 84 | ||
| XM_006241085.2 | XP_006241147.1 | 1549 | 84 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Cyp4f17
Genomic Map