Assay Details
| Gene Symbol: Mbd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108735.1 | NP_001102205.1 | 5 - 6 | 979 | 64 | |
| XM_006240965.1 | XP_006241027.1 | 5 - 6 | 973 | 64 | ||
| XM_006240966.1 | XP_006241028.1 | 5 - 6 | 872 | 64 | ||
| XM_008765119.1 | XP_008763341.1 | 6 - 7 | 851 | 64 | ||
| XM_008765120.1 | XP_008763342.1 | 5 - 6 | 791 | 64 | ||
| XM_008765121.1 | XP_008763343.1 | 6 - 7 | 775 | 64 | ||
| XM_008765122.1 | XP_008763344.1 | 5 - 6 | 971 | 64 | ||
| GenBank mRNA | BC167041.1 | - | 5 - 6 | 746 | 64 | |
| FQ216718.1 | - | 4 - 5 | 487 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mbd3
Genomic Map