Assay Details
| Gene Symbol: Lmnb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006241035.2 | XP_006241097.1 | 4 - 5 | 1998 | 79 | |
| XM_008765148.1 | XP_008763370.1 | 8 - 9 | 1999 | 79 | ||
| XM_008765149.1 | XP_008763371.1 | 8 - 9 | 2270 | 79 | ||
| XM_216850.8 | XP_216850.6 | 7 - 8 | 2411 | 79 | ||
| GenBank mRNA | FQ212316.1 | - | 7 - 8 | 1235 | 79 | |
More Information
Set Membership: |
|

Lmnb2
Genomic Map