Assay Details
| Gene Symbol: Bri3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009604.1 | NP_001009604.1 | 1 - 2 | 243 | 138 | |
| GenBank mRNA | BC087646.1 | - | 1 - 2 | 243 | 7311234 | 138 |
More Information
Updated Mapping Information
Set Membership: |
|

Bri3
Genomic Map