Assay Details
| Gene Symbol: Ptcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109665.1 | NP_001103135.1 | 6 - 7 | 1709 | 71 | |
| XM_006248871.1 | XP_006248933.1 | 7 - 8 | 1840 | 71 | ||
| XM_006248873.1 | XP_006248935.1 | 6 - 7 | 1699 | 71 | ||
| GenBank mRNA | BC129116.1 | - | 7 - 8 | 1749 | 7113830 | 71 |
More Information
Updated Mapping Information
Set Membership: |
|

Ptcd1
Genomic Map