Assay Details
| Gene Symbol: Cyp3a62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024232.1 | NP_001019403.1 | 1 - 2 | 258 | 133 | |
| GenBank mRNA | AB084894.1 | - | 1 - 2 | 258 | 133 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cyp3a62
Genomic Map