Assay Details
| Gene Symbol: Dhrsx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105914.1 | NP_001099384.1 | 2 - 3 | 204 | 54 | |
| XM_006248993.2 | XP_006249055.1 | 2 - 3 | 255 | 54 | ||
| XM_006248995.2 | XP_006249057.1 | 2 - 3 | 256 | 54 | ||
| XM_006248997.1 | XP_006249059.1 | 2 - 3 | 499 | 54 | ||
| GenBank mRNA | BC167053.1 | - | 2 - 3 | 183 | 7110020 | 54 |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006248997.1 transcripts.
Set Membership: |
|

Dhrsx
Genomic Map