Assay Details
| Gene Symbol: Arpc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031146.2 | NP_112408.1 | 3 - 4 | 314 | 81 | |
| GenBank mRNA | AF315378.1 | - | 2 - 3 | 199 | 81 | |
| BC059131.1 | - | 3 - 4 | 294 | 6921520 | 81 | |
| DY310102.1 | - | 3 - 4 | 314 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arpc1a
Genomic Map