Assay Details
| Gene Symbol: Arpc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031146.2 | NP_112408.1 | 5 - 6 | 648 | 67 | |
| GenBank mRNA | AF315378.1 | - | 4 - 5 | 533 | 67 | |
| CO396710.1 | - | 116 | 67 | |||
| DY310102.1 | - | 5 - 6 | 648 | 67 | ||
| FQ214345.1 | - | 5 - 6 | 622 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arpc1a
Genomic Map