Assay Details
| Gene Symbol: Arpc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031146.2 | NP_112408.1 | 8 - 9 | 1131 | 95 | |
| GenBank mRNA | AF315378.1 | - | 7 - 8 | 1016 | 95 | |
| FQ214345.1 | - | 8 - 9 | 1107 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arpc1a
Genomic Map